Methyl hydrogen phthalate, 97%, Thermo Scientific Chemicals
Methyl hydrogen phthalate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl hydrogen phthalate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14780.09
also known as A14780-09
Price (USD)/ Each
58.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
58.10
Add to cart
Methyl hydrogen phthalate, 97%, Thermo Scientific Chemicals
Catalog numberA14780.09
Price (USD)/ Each
58.10
-
Add to cart
Chemical Identifiers
CAS4376-18-5
IUPAC Name2-(methoxycarbonyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyFNJSWIPFHMKRAT-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to pale cream
Methyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. ML Bender.; F Chloupek.; MC Neveu. Intramolecular Catalysis of Hydrolytic Reactions. III. Intramolecular Catalysis by Carboxylate Ion in the Hydrolysis of Methyl Hydrogen Phthalate1, 2. J. Am. Chem. Soc.1958, 80 (20), 5384-5387.
  2. V Rao.; PV Ashokan.; MH Shridhar. Studies on the compatibility and specific interaction in cellulose acetate hydrogen phthalate(CAP) and poly methyl methacrylate (PMMA) blend. Polymer.1999, 40 (25), 7167-7171.
  3. Undergoes a modified Curtius rearrangement via a mixed anhydride. The resulting isocyanate has been cyclized with, e.g. alanine to give chiral quinazoline-2,4-dione derivatives: Heterocycles, 36, 1305 (1993):