Thermo Scientific Chemicals

2-(Trifluoromethyl)phenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: A14781.03
1 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)phenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: A14781.03
1 g, Each
Quantity
Catalog number: A14781.03
also known as A14781-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
365-34-4
IUPAC Name
1-phenyl-2-(trifluoromethyl)hydrazine
Molecular Formula
C7H7F3N2
InChI Key
LJORWWOXCXNLOT-UHFFFAOYSA-N
SMILES
FC(F)(F)NNC1=CC=CC=C1
Appearance (Color)
White to yellow or cream to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
58.0-67.0°C

Description

2-(Trifluoromethyl)phenylhydrazine is used as an intermediate in organic syntheses. It is also ued as a fine chemical intermediate, GC derivatizing agent, coupling reagent and N- protecting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenylhydrazine is used as an intermediate in organic syntheses. It is also ued as a fine chemical intermediate, GC derivatizing agent, coupling reagent and N- protecting agent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text