Heptafluoro-1-iodopropane, 94%, stab. with copper, Thermo Scientific Chemicals
Heptafluoro-1-iodopropane, 94%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Heptafluoro-1-iodopropane, 94%, stab. with copper, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A14784.14
also known as A14784-14
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
Add to cart
Heptafluoro-1-iodopropane, 94%, stab. with copper, Thermo Scientific Chemicals
Catalog numberA14784.14
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS754-34-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow or pale purple
FormLiquid
Assay (GC)≥92.0%
Refractive Index1.3255-1.3325 @ 20°C
StabilizerCopper powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only