Thermo Scientific Chemicals

2-Chlorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A14785.22
100 g, Each
Thermo Scientific Chemicals

2-Chlorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A14785.22
100 g, Each
Quantity
Catalog number: A14785.22
also known as A14785-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
609-65-4
IUPAC Name
2-chlorobenzoyl chloride
Molecular Formula
C7H4Cl2O
InChI Key
ONIKNECPXCLUHT-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=CC=C1Cl
Form
Liquid
Refractive Index
1.5705-1.5755 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow

Description

2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoyl chloride, is used in organic synthesis. It is used as a pharmaceutical intermediate for eg as an intermediate of clotrimazolc, preparation of p-chlorobenzoic acid and production of trichloro-acaricide.

Solubility
Soluble in acetone, ether, and alcohol. Solubility in water it decomposes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong bases, alcohols, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text