Thermo Scientific Chemicals

2-Acetylnaphthalene, 99%, Thermo Scientific Chemicals

Catalog number: A14793.18
50 g, Each
Thermo Scientific Chemicals

2-Acetylnaphthalene, 99%, Thermo Scientific Chemicals

Catalog number: A14793.18
50 g, Each
Quantity
Catalog number: A14793.18
also known as A14793-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-08-3
IUPAC Name
1-(naphthalen-2-yl)ethan-1-one
Molecular Formula
C12H10O
InChI Key
XSAYZAUNJMRRIR-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C2C=CC=CC2=C1
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder or lumps or chunks
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
50.0-57.0?C

Description

2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

Solubility
Soluble in ethanol, acetone and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text