3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,6-Dichloropyridazine is used as an intermediate in synthetic chemistry, especially in active pharmaceutical ingredients. It is used in the preparation of alfa-(halohetaroyl)-2-azahetarylacetonitriles.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Neubert, T. D.; Schmidt, Y.; Conroy, E.; Stamos, D. Radical Mediated C-H Functionalization of 3,6-Dichloropyridazine: Efficient Access to Novel Tetrahydropyridopyridazines. Org. Lett. 2015, 17 (10), 2362-2365.
Hübscher, J.; Gruber, T.; Seichter, W.; Kortus, J.; Weber, E. Intramolecular fixation of t-butyl groups in thiolactim ethers influencing molecular conformation and the packing behavior. J. Mol. Struct. 2015, 1091, 88-97.