Thermo Scientific Chemicals

3,5-Dimethoxy-4-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A14801.22
100 g, Each
Thermo Scientific Chemicals

3,5-Dimethoxy-4-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: A14801.22
100 g, Each
Quantity
Catalog number: A14801.22
also known as A14801-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61040-81-1
IUPAC Name
3,5-dimethoxy-4-methylbenzoate
Molecular Formula
C10H11O4
InChI Key
QIBMVRYNEXOCCF-UHFFFAOYSA-M
SMILES
COC1=CC(=CC(OC)=C1C)C([O-])=O
Appearance (Color)
Pale cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Silylated GC)
≥96.0%
Melting Point (clear melt)
213.5-222.5?C

Description

3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxy-4-methylbenzoic acid undergoes reduction in the presence of sodium in ethanol to prepare 1,4-dihydro-3,5-dimethoxy- kmethylbenzoic acid.

Solubility
Soluble in liquid ammonia.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text