Thermo Scientific Chemicals

1,2-Dibromo-4,5-difluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A14804.22
100 g, Each
Thermo Scientific Chemicals

1,2-Dibromo-4,5-difluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A14804.22
100 g, Each
Quantity
Catalog number: A14804.22
also known as A14804-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64695-78-9
IUPAC Name
1,2-dibromo-4,5-difluorobenzene
Molecular Formula
C6H2Br2F2
InChI Key
JTEZQWOKRHOKDG-UHFFFAOYSA-N
SMILES
FC1=CC(Br)=C(Br)C=C1F
Appearance (Color)
White to pale yellow or pale brown
Form
Crystalline or fused solid, clear as melt
Assay (GC)
> 97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text