Thermo Scientific Chemicals

Sodium 4-hydroxybenzenesulfonate dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: A14808.0E
2500 g, Each
Thermo Scientific Chemicals

Sodium 4-hydroxybenzenesulfonate dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: A14808.0E
2500 g, Each
Quantity
Catalog number: A14808.0E
also known as A14808-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10580-19-5
IUPAC Name
4-hydroxybenzene-1-sulfonate
Molecular Formula
C6H5O4S
InChI Key
FEPBITJSIHRMRT-UHFFFAOYSA-M
SMILES
OC1=CC=C(C=C1)S([O-])(=O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (ex Sulphated Ash)
≥96.0% (as anhydrous, dry wt. basis)
Water Content (Karl Fischer Titration)
14-17% (1.77 - 2.23 waters)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text