5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.
Solubility
Soluble in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
RUO – Research Use Only
Satoru Ito .; Masakatsu Matsumoto. Ferric salt-catalyzed oxygenation of cycloalkanones to oxo esters by molecular oxygen. J. Org. Chem. 1983, 48, (7), 1133-1135.
Timothy L. Macdonald.; Robert Dolan. Regiospecific 2.beta.-chloro-3-tropinone preparation. A synthesis of tropinone and pseudopelletierine. J. Org. Chem. 1979, 44, (26), 4973-4976.