Thermo Scientific Chemicals

Xanthone, 99%, Thermo Scientific Chemicals

Catalog number: A14812.14
25 g, Each
Thermo Scientific Chemicals

Xanthone, 99%, Thermo Scientific Chemicals

Catalog number: A14812.14
25 g, Each
Quantity
Catalog number: A14812.14
also known as A14812-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90-47-1
IUPAC Name
9H-xanthen-9-one
Molecular Formula
C13H8O2
InChI Key
JNELGWHKGNBSMD-UHFFFAOYSA-N
SMILES
O=C1C2=CC=CC=C2OC2=CC=CC=C12
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale brown or pale grey
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
172.5-178.5?C

Description

Xanthone is used predominantly as oxygenated polycyclic aromatic compounds, and as a vasorelaxant. It can be used as a fluorescent agent, as well as an intermediate in organic synthesis and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xanthone is used predominantly as oxygenated polycyclic aromatic compounds, and as a vasorelaxant. It can be used as a fluorescent agent, as well as an intermediate in organic synthesis and chemical research.

Solubility
It is Soluble in water (partly), chloroform, hot Toluene, ether (slightly), and hot alcohol.

Notes
Stable under normal conditions. Incompatible to strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text