Thermo Scientific Chemicals

Tris(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: A14813.03
1 g, Each
Thermo Scientific Chemicals

Tris(4-fluorophenyl)phosphine, 98%, Thermo Scientific Chemicals

Catalog number: A14813.03
1 g, Each
Quantity
Catalog number: A14813.03
also known as A14813-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18437-78-0
IUPAC Name
tris(4-fluorophenyl)phosphane
Molecular Formula
C18H12F3P
InChI Key
GEPJPYNDFSOARB-UHFFFAOYSA-N
SMILES
FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
77.0-84.0?C

Description

Tris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text