Thermo Scientific Chemicals

N-(Ethoxycarbonyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: A14822.14
25 g, Each
Thermo Scientific Chemicals

N-(Ethoxycarbonyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: A14822.14
25 g, Each
Quantity
Catalog number: A14822.14
also known as A14822-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
22509-74-6
IUPAC Name
ethyl 1,3-dioxo-2,3-dihydro-1H-isoindole-2-carboxylate
Molecular Formula
C11H9NO4
InChI Key
VRHAQNTWKSVEEC-UHFFFAOYSA-N
SMILES
CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O
Melting Point (clear melt)
76.0-86.0?C
Form
Powder
Appearance (Color)
White
Assay (GC)
≥96.0%

Description

N-(Ethoxycarbonyl)phthalimide is used to protect amine functional groups. Reagent for the conversion of amines and amino acids to their phthaloyl derivatives under mild conditions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Ethoxycarbonyl)phthalimide is used to protect amine functional groups. Reagent for the conversion of amines and amino acids to their phthaloyl derivatives under mild conditions

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text