Thermo Scientific Chemicals

Elaidic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14832.03
1 g, Each
Thermo Scientific Chemicals

Elaidic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14832.03
1 g, Each
Quantity
Catalog number: A14832.03
also known as A14832-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-79-8
IUPAC Name
(9E)-octadec-9-enoic acid
Molecular Formula
C18H34O2
InChI Key
ZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILES
CCCCCCCCC=CCCCCCCCC(O)=O
Appearance (Color)
White to pale yellow
Form
Powder or crystalline powder or flakes
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
41-46?C
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In human platelets incubated with arachidonic acid, elaidic acid inhibits HHT and HETE formation while inducing prostaglandin and thromboxane synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text