p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Terphenyl is a specialty material, and may be used in ionized radiation detectors, nonpolar laser dye, and single molecule optical probe of scanning near-field microscopy.
Solubility
Insoluble in water. Soluble in hot benzene. Very soluble in hot ethyl alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only
Boens N.; Qin W, Basarić N.; Hofkens J.; Ameloot M.; Pouget J.; Lefèvre JP.etal. Fluorescence lifetime standards for time and frequency domain fluorescence spectroscopy. Analytical chemistry . 2007, 79, (3), 2137-2149 .•JK Liu .; L Hu .; ZJ Dong .; Q Hu. DPPH Radical Scavenging Activity of Ten Natural p-Terphenyl Derivatives Obtained from Three Edible Mushrooms Indigenous to China. Chemistry & biodiversity. 2004, 1, (4), 601-605.