Thermo Scientific Chemicals

n-Dodecane, ≥99%, Thermo Scientific Chemicals

Catalog number: A14834.AP
500 mL, Each
Thermo Scientific Chemicals

n-Dodecane, ≥99%, Thermo Scientific Chemicals

Catalog number: A14834.AP
500 mL, Each
Quantity
Catalog number: A14834.AP
also known as A14834-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-40-3
IUPAC Name
dodecane
Molecular Formula
C12H26
InChI Key
SNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC
Assay from Supplier's CofA
≥98.5% (US-sourced material)
Comment
Material sourced in UK and US
Identification (FTIR)
Conforms (UK-sourced material)
Appearance (Color)
Clear colorless
Form
Liquid

Description

n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Dodecane is used as a solvent and a distillation chaser. It finds application as a diluent for tributyl phosphate (TBP) in reprocessing plants and as a possible surrogate for kerosene-based fuels in jet. It is an active component of scintillator as well as used in lubricants and greases.

Solubility
Miscible with ethyl alcohol, ethyl ether, acetone, chloroform and carbon tetrachloride. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text