Thermo Scientific Chemicals

2-Bromothiazole, 99%, Thermo Scientific Chemicals

Catalog number: A14838.14
25 g, Each
Thermo Scientific Chemicals

2-Bromothiazole, 99%, Thermo Scientific Chemicals

Catalog number: A14838.14
25 g, Each
Quantity
Catalog number: A14838.14
also known as A14838-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3034-53-5
IUPAC Name
2-bromo-1,3-thiazole
Molecular Formula
C3H2BrNS
InChI Key
RXNZFHIEDZEUQM-UHFFFAOYSA-N
SMILES
BrC1=NC=CS1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5915-1.5945 @ 20?C

Description

2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text