Thermo Scientific Chemicals

2,6-Dichlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14839.09
10 g, Each
Thermo Scientific Chemicals

2,6-Dichlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A14839.09
10 g, Each
Quantity
Catalog number: A14839.09
also known as A14839-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4659-45-4
IUPAC Name
2,6-dichlorobenzoyl chloride
Molecular Formula
C7H3Cl3O
InChI Key
JBLIDPPHFGWTKU-UHFFFAOYSA-N
SMILES
ClC(=O)C1=C(Cl)C=CC=C1Cl
Refractive Index
1.5580-1.5620 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

2,6-Dichlorobenzoyl chloride acts as an important intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is employed in the substrate activity screening method for the rapid development of novel substrates and their conversion into non-peptidic inhibitors of Cys and Ser proteases. It is also used in the preparation of 1-acyliridoles and (-)-aspicilin. In addition to this, it undergoes esterification of (fluoren-9-ylmethoxy)carbonyl (Fmoc)-amino acids to 4-alkoxybenzyl alcohol polystyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorobenzoyl chloride acts as an important intermediate in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is employed in the substrate activity screening method for the rapid development of novel substrates and their conversion into non-peptidic inhibitors of Cys and Ser proteases. It is also used in the preparation of 1-acyliridoles and (-)-aspicilin. In addition to this, it undergoes esterification of (fluoren-9-ylmethoxy)carbonyl (Fmoc)-amino acids to 4-alkoxybenzyl alcohol polystyrene.

Solubility
Miscible with alcohol, ether and acetone. Slightly miscible with heptane. Immiscible with water.

Notes
Moisture sensitive. Incompatible with alcohols, oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text