5-Bromo-2-furoic acid, 99%, Thermo Scientific Chemicals
5-Bromo-2-furoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-furoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A14840.18
also known as A14840-18
Price (USD)/ Each
80.30
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
80.30
Add to cart
5-Bromo-2-furoic acid, 99%, Thermo Scientific Chemicals
Catalog numberA14840.18
Price (USD)/ Each
80.30
-
Add to cart
Chemical Identifiers
CAS585-70-6
IUPAC Name5-bromofuran-2-carboxylic acid
Molecular FormulaC5H3BrO3
InChI KeyYVTQHZDUDUCGRD-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Br)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (HPLC)≥98.5%
Melting Point (clear melt)187.0-193.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only