Thermo Scientific Chemicals

3-Methyl-1-phenyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog number: A14841.03
1 g, Each
Thermo Scientific Chemicals

3-Methyl-1-phenyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog number: A14841.03
1 g, Each
Quantity
Catalog number: A14841.03
also known as A14841-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1128-54-7
IUPAC Name
3-methyl-1-phenyl-1H-pyrazole
Molecular Formula
C10H10N2
InChI Key
RJXLUGSJEMSDPK-UHFFFAOYSA-N
SMILES
CC1=NN(C=C1)C1=CC=CC=C1
Appearance (Color)
White to cream or pale yellow
Form
Crystals or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
33-37?C

Description

3-Methyl-1-phenyl-1H-pyrazole is used to produce 3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde at the temperature of 90-100°C with reagent phosphorus oxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-1H-pyrazole is used to produce 3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde at the temperature of 90-100°C with reagent phosphorus oxychloride.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text