3-Methyl-1-phenyl-1H-pyrazole is used to produce 3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde at the temperature of 90-100°C with reagent phosphorus oxychloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methyl-1-phenyl-1H-pyrazole is used to produce 3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde at the temperature of 90-100°C with reagent phosphorus oxychloride.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only