Thermo Scientific Chemicals

4-Benzyloxyphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A14843.14
25 g, Each
Thermo Scientific Chemicals

4-Benzyloxyphenol, 98+%, Thermo Scientific Chemicals

Catalog number: A14843.14
25 g, Each
Quantity
Catalog number: A14843.14
also known as A14843-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-16-2
IUPAC Name
4-(benzyloxy)phenol
Molecular Formula
C13H12O2
InChI Key
VYQNWZOUAUKGHI-UHFFFAOYSA-N
SMILES
OC1=CC=C(OCC2=CC=CC=C2)C=C1
Appearance (Color)
White to cream to pale brown to pale gray
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text