Thermo Scientific Chemicals

Isophorone, 97%, Thermo Scientific Chemicals

Catalog number: A14844.0F
2500 mL, Each
Thermo Scientific Chemicals

Isophorone, 97%, Thermo Scientific Chemicals

Catalog number: A14844.0F
2500 mL, Each
Quantity
Catalog number: A14844.0F
also known as A14844-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-59-1
IUPAC Name
3,5,5-trimethylcyclohex-2-en-1-one
Molecular Formula
C9H14O
InChI Key
HJOVHMDZYOCNQW-UHFFFAOYSA-N
SMILES
CC1=CC(=O)CC(C)(C)C1
Refractive Index
1.4750-1.4800 @ 20°C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings

Solubility
Soluble in water (12g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text