Isophorone, 97%, Thermo Scientific Chemicals
Isophorone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isophorone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
2500 mL
Catalog number A14844.AK
also known as A14844-AK
Price (USD)/ Each
24.10
-
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)/ Each
24.10
Add to cart
Isophorone, 97%, Thermo Scientific Chemicals
Catalog numberA14844.AK
Price (USD)/ Each
24.10
-
Add to cart
Chemical Identifiers
CAS78-59-1
IUPAC Name3,5,5-trimethylcyclohex-2-en-1-one
Molecular FormulaC9H14O
InChI KeyHJOVHMDZYOCNQW-UHFFFAOYSA-N
SMILESCC1=CC(=O)CC(C)(C)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.4750-1.4800 @ 20°C
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone is used as a solvent and as an intermediate in organic synthesis. Isophorone also occurs naturally in cranberries. Isophorone is used as a solvent in some printing inks, paints, lacquers, adhesives, copolymers, coatings, finishings

Solubility
Soluble in water (12g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. M. S. Kharasch.; P. O. Tawney. Factors Determining the Course and Mechanisms of Grignard Reactions. II. The Effect of Metallic Compounds on the Reaction between Isophorone and Methylmagnesium Bromide. J. Am. Chem. Soc. 1941, 63, (9), 2308-2316.
  2. N.S. Zarghami.; D.E. Heinz. Monoterpene aldehydes and isophorone-related compounds of saffron. Phytochemistry. 1971, 10, (11), 2755-2761.