Thermo Scientific Chemicals

Thioanisole, 99%, Thermo Scientific Chemicals

Catalog number: A14846.0E
2500 g, Each
Thermo Scientific Chemicals

Thioanisole, 99%, Thermo Scientific Chemicals

Catalog number: A14846.0E
2500 g, Each
Quantity
Catalog number: A14846.0E
also known as A14846-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-68-5
IUPAC Name
(methylsulfanyl)benzene
Molecular Formula
C7H8S
InChI Key
HNKJADCVZUBCPG-UHFFFAOYSA-N
SMILES
CSC1=CC=CC=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5850-1.5880 @ 20?C
Assay (GC)
≥98.5%

Description

Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text