3-Cyanopyridine, 98%, Thermo Scientific Chemicals
3-Cyanopyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Cyanopyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A14850.36
also known as A14850-36
Price (USD)/ Each
104.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
104.00
Add to cart
3-Cyanopyridine, 98%, Thermo Scientific Chemicals
Catalog numberA14850.36
Price (USD)/ Each
104.00
-
Add to cart
Chemical Identifiers
CAS100-54-9
IUPAC Namepyridine-3-carbonitrile
Molecular FormulaC6H4N2
InChI KeyGZPHSAQLYPIAIN-UHFFFAOYSA-N
SMILESN#CC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)47.0-53.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only