Thermo Scientific Chemicals

Pyrrolidine, 99%, Thermo Scientific Chemicals

Catalog number: A14852.AP
500 mL, Each
Thermo Scientific Chemicals

Pyrrolidine, 99%, Thermo Scientific Chemicals

Catalog number: A14852.AP
500 mL, Each
Quantity
Catalog number: A14852.AP
also known as A14852-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123-75-1
IUPAC Name
hydrogen pyrrolidine chloride
Molecular Formula
C4H10ClN
InChI Key
FCLZCOCSZQNREK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].C1CCNC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Color (APHA)
≤50 (U.S. specification)
Assay (GC)
≥98.5%
Comment
Purchased in U.K. and U.S.

Description

Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

Solubility
Miscible with alcohol, ether, chloroform and water.

Notes
Store in cool place. Incompatible with acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text