Thermo Scientific Chemicals

2-Chloropyridine, 99%, Thermo Scientific Chemicals

Catalog number: A14866.22
100 g, Each
Thermo Scientific Chemicals

2-Chloropyridine, 99%, Thermo Scientific Chemicals

Catalog number: A14866.22
100 g, Each
Quantity
Catalog number: A14866.22
also known as A14866-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-09-1
IUPAC Name
2-chloropyridine
Molecular Formula
C5H4ClN
InChI Key
OKDGRDCXVWSXDC-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=N1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Refractive Index
1.5315-1.5345 @ 20?C
Assay (GC)
≥98.5%

Description

2- Chloropyridine is used as an intermediate chemical in the production of various pyrithione, for use in cosmetics, personal care products and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2- Chloropyridine is used as an intermediate chemical in the production of various pyrithione, for use in cosmetics, personal care products and pharmaceutical products.

Solubility
Soluble in water (27g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text