Tri-n-butylamine, 98%, Thermo Scientific Chemicals
Tri-n-butylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-butylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A14868.0D
also known as A14868-0D
Price (USD)/ Each
303.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
357.00 
Save 53.35 (15%)
-
Add to cart
Quantity:
10,000 mL
Request bulk or custom format
Price (USD)/ Each
303.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
357.00 
Save 53.35 (15%)
Add to cart
Tri-n-butylamine, 98%, Thermo Scientific Chemicals
Catalog numberA14868.0D
Price (USD)/ Each
303.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
357.00 
Save 53.35 (15%)
-
Add to cart
Chemical Identifiers
CAS102-82-9
IUPAC Nametributylamine
Molecular FormulaC12H27N
InChI KeyIMFACGCPASFAPR-UHFFFAOYSA-N
SMILESCCCCN(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5% (U.S. sourced material)
Refractive Index1.4270-1.4310 @ 20°C (non-U.S. sourced material)
Assay (GC)≥97.5% (non-U.S. sourced material)
Identification (FTIR)Conforms (non-U.S. sourced material)
Appearance (Color)Clear colorless to pale yellow
View more
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

Solubility
Miscible with acetone, ethanol, ethyl ether and most organic solvents. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Nakajima, M.; Fava, E.; Loescher, S.; Jiang, Z.; Rueping, M. Photoredox-Catalyzed Reductive Coupling of Aldehydes, Ketones, and Imines with Visible Light. Angew. Chem. Int. Ed. 2015, 54 (30), 8828-8832.
  2. Sarac, I.; Meier, C. Efficient Automated Solid-Phase Synthesis of DNA and RNA 5'-Triphosphates. Chem. Eur. J. 2015, 21 (46), 16421-16426.