Thermo Scientific Chemicals

3-Amino-1H-1,2,4-triazole, 96%, Thermo Scientific Chemicals

Catalog number: A14871.0B
1000 g, Each
Thermo Scientific Chemicals

3-Amino-1H-1,2,4-triazole, 96%, Thermo Scientific Chemicals

Catalog number: A14871.0B
1000 g, Each
Quantity
Catalog number: A14871.0B
also known as A14871-0B
Price (USD)
Have Questions?

Chemical Identifiers

CAS
61-82-5
IUPAC Name
1H-1,2,4-triazol-5-amine
Molecular Formula
C2H4N4
InChI Key
KLSJWNVTNUYHDU-UHFFFAOYSA-N
SMILES
NC1=NC=NN1
Appearance (Color)
White to cream
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0%
Assay (HPLC)
≥95.0%
Melting Point (clear melt)
145.0-158.0?C
Form
Powder

Description

3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.

Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text