Thermo Scientific Chemicals

4-Bromophenol, 99%, Thermo Scientific Chemicals

Catalog number: A14873.0E
2500 g, Each
Thermo Scientific Chemicals

4-Bromophenol, 99%, Thermo Scientific Chemicals

Catalog number: A14873.0E
2500 g, Each
Quantity
Catalog number: A14873.0E
also known as A14873-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-41-2
IUPAC Name
4-bromophenol
Molecular Formula
C6H5BrO
InChI Key
GZFGOTFRPZRKDS-UHFFFAOYSA-N
SMILES
OC1=CC=C(Br)C=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream or pink
Melting Point (clear melt)
62.0-68.0?C

Description

4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.

Solubility
It is insoluble in water. It is soluble in 5%ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Store under inert gas. Air sensitive. Keep away from Oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text