4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromophenol is used as an intermediate in medicine grade. It is also used as a phenol that is used in Stille reactions. It is used in the intermediate of organic synthesis, flame retardant and pharmaceutical industry.
Solubility
It is insoluble in water. It is soluble in 5%ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Store under inert gas. Air sensitive. Keep away from Oxidizing agents.
RUO – Research Use Only
N.W. Larsen. Microwave spectra and internal rotation of 4-fluorophenol, 4-chlorophenol and 4-bromophenol. Journal of Molecular Structure. 1986, 144, (1), 83-99.