Thermo Scientific Chemicals

Triphenylmethyl mercaptan, 98+%, Thermo Scientific Chemicals

Catalog number: A14888.06
5 g, Each
Thermo Scientific Chemicals

Triphenylmethyl mercaptan, 98+%, Thermo Scientific Chemicals

Catalog number: A14888.06
5 g, Each
Quantity
Catalog number: A14888.06
also known as A14888-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3695-77-0
IUPAC Name
triphenylmethanethiol
Molecular Formula
C19H16S
InChI Key
JQZIKLPHXXBMCA-UHFFFAOYSA-N
SMILES
SC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to cream or pale yellow
Form
Crystalline powder
Melting Point (clear melt)
102-109?C
Assay (HPLC)
≥98.0%
Identification (FTIR)
Conforms

Description

Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text