Thermo Scientific Chemicals

1-Bromoadamantane, 99%, Thermo Scientific Chemicals

Catalog number: A14893.14
25 g, Each
Thermo Scientific Chemicals

1-Bromoadamantane, 99%, Thermo Scientific Chemicals

Catalog number: A14893.14
25 g, Each
Quantity
Catalog number: A14893.14
also known as A14893-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
768-90-1
IUPAC Name
1-bromoadamantane
Molecular Formula
C10H15Br
InChI Key
VQHPRVYDKRESCL-UHFFFAOYSA-N
SMILES
BrC12CC3CC(CC(C3)C1)C2
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
115.0-121.0?C

Description

1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoadamantane is an important intermediate, used in synthesizing medicines and new materials etc.

Solubility
It is soluble in organic solvents and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep away from Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text