Thermo Scientific Chemicals

Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals

Catalog number: A14894.14
25 g, Each
Thermo Scientific Chemicals

Vitamin B12, 98+% (dry wt basis), Thermo Scientific Chemicals

Catalog number: A14894.14
25 g, Each
Quantity
Catalog number: A14894.14
also known as A14894-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68-19-9
IUPAC Name
(10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-1-cyano-46-hydroxy-12-(hydroxymethyl)-5,6,17,23,28,31,31,36,38,41,42-undecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
Molecular Formula
C63H88CoN14O14P
InChI Key
YUWVGNQGDAPKCX-BVWPOUIRNA-L
SMILES
[C@@H]1(C2=CC3=[N+]4C(C(C3(C)C)CCC(N)=O)=C(C3=[N+]5C6C7(N8C(C([C@@]7(C)CC(N)=O)CCC(N)=O)=C(C(=[N+]2[Co+]458([N+]2=CN(C4=CC(=C(C)C=C24)C)[C@@]2([H])O[C@@H]([C@]([H])(OP([O-])(=O)OC(C)CNC(=O)CCC3(C)[C@@H]6CC(N)=O)C2O)CO)C#N)C1(C)CC(N)=O)C)C)C)CCC(N)=O
Form
crystals or crystalline powder
Assay (unspecified)
> 98.0% (on dried material)
Loss on Drying
≤ 5.0%
Appearance (Color)
Dark red to red-purple

Description

Vitamin B12 is used as a nutritional supplement and stabilizer in foods. It is also used as a medicine and animal feed supplement. It plays an important role in the synthesis of myelin. It is also used as an antipernicious anemia factor. Further, it is used to treat cyanide poisoning and hereditary deficiency of transcobalamin II.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text