(Chloromethyl)dimethylchlorosilane, 98%, Thermo Scientific Chemicals
(Chloromethyl)dimethylchlorosilane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Chloromethyl)dimethylchlorosilane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
100 g
Catalog number A14899.14
also known as A14899-14
Price (USD)/ Each
48.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
48.60
Add to cart
(Chloromethyl)dimethylchlorosilane, 98%, Thermo Scientific Chemicals
Catalog numberA14899.14
Price (USD)/ Each
48.60
-
Add to cart
Chemical Identifiers
CAS1719-57-9
IUPAC Namechloro(chloromethyl)dimethylsilane
Molecular FormulaC3H8Cl2Si
InChI KeyITKVLPYNJQOCPW-UHFFFAOYSA-N
SMILESC[Si](C)(Cl)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
CommentSpecification differs for U.S. and non-U.S. material where indicated
Refractive Index1.4350-1.4390 @ 20°C (non-U.S. specification)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only