4-Chloromethyl-2-methylthiazole hydrochloride, 98%, Thermo Scientific Chemicals
4-Chloromethyl-2-methylthiazole hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloromethyl-2-methylthiazole hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14902.06
also known as A14902-06
Price (USD)/ Each
162.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
162.00
Add to cart
4-Chloromethyl-2-methylthiazole hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberA14902.06
Price (USD)/ Each
162.00
-
Add to cart
Chemical Identifiers
CAS77470-53-2
IUPAC Name4-(chloromethyl)-2-methyl-1,3-thiazole hydrochloride
Molecular FormulaC5H7Cl2NS
InChI KeyYGKDISJLDVGNOR-UHFFFAOYSA-N
SMILESCl.CC1=NC(CCl)=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)to pale gray
FormPowder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only