Thermo Scientific Chemicals

2,2,2-Trifluoroethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A14904.22
100 g, Each
Thermo Scientific Chemicals

2,2,2-Trifluoroethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A14904.22
100 g, Each
Quantity
Catalog number: A14904.22
also known as A14904-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
373-88-6
Appearance (Color)
White to yellow
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Assay (Freebase GC)
≥97.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1%

Description

2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethylamine hydrochloride, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff

Solubility
It is Soluble in 0.7 to water, but soluble in ethanol and chloroform, slightly soluble in benzene, hardly soluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away from strong oxidizing agents, acids, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text