Thermo Scientific Chemicals

Adenine, 99%

Catalog number: A14906.36
500 g, Each
Thermo Scientific Chemicals

Adenine, 99%

Catalog number: A14906.36
500 g, Each
Quantity
Catalog number: A14906.36
also known as A14906-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
73-24-5
IUPAC Name
7H-purin-6-amine
Molecular Formula
C5H5N5
InChI Key
GFFGJBXGBJISGV-UHFFFAOYSA-N
SMILES
NC1=C2NC=NC2=NC=N1
Appearance (Color)
White to pale cream
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

Solubility
Soluble in hot water, methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text