Thermo Scientific Chemicals

Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: A14907.14
25 g, Each
Thermo Scientific Chemicals

Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: A14907.14
25 g, Each
Quantity
Catalog number: A14907.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
375-22-4
IUPAC Name
heptafluorobutanoic acid
Molecular Formula
C4HF7O2
InChI Key
YPJUNDFVDDCYIH-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)C(F)(F)C(F)(F)F
Appearance (Color)
Clear colorless to pale yellow or pale pink
Form
Liquid
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.

Solubility
Miscible with water, toluene, ether, ethanol and benzene.       

Notes
Hygroscopic. Light sensitive. Incompatible withbases, oxidizing agents and reducing agents..   
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text