Thermo Scientific Chemicals

3-Phenylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14908.22
100 g, Each
Thermo Scientific Chemicals

3-Phenylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14908.22
100 g, Each
Quantity
Catalog number: A14908.22
also known as A14908-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
501-52-0
IUPAC Name
3-phenylpropanoic acid
Molecular Formula
C9H10O2
InChI Key
XMIIGOLPHOKFCH-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=CC=CC=C1
Appearance (Color)
White to cream to yellow to pale brown
Melting Point (clear melt)
46-52°C
Assay from Suppliers CofA
≥98.5%
Form
Crystalline powder or crystals or fused solid

Description

Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A?s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A′s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text