4-Cyanobenzaldehyde, 98+%, Thermo Scientific Chemicals
4-Cyanobenzaldehyde, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyanobenzaldehyde, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A14914.14
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
Add to cart
4-Cyanobenzaldehyde, 98+%, Thermo Scientific Chemicals
Catalog numberA14914.14
Price (USD)/ Each
170.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
201.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS105-07-7
IUPAC Name4-formylbenzonitrile
Molecular FormulaC8H5NO
InChI KeyWZWIQYMTQZCSKI-UHFFFAOYSA-N
SMILESO=CC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
FormCrystals or powder or crystalline powder or lumps
Free acid (titration)≤1%
Identification (FTIR)Conforms
Melting Point (clear melt)97.0-103.0?C
View more
4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.

Solubility
It is soluble in water, ethanol, ether and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.
RUO – Research Use Only
Joelle Vidal .; Laure Guy .; Sebastien Sterin .; Andre Collet. Electrophilic amination: preparation and use of N-Boc-3-(4-cyanophenyl)oxaziridine, a new reagent that transfers a N-Boc group to N- and C-nucleophiles.. J. Org. Chem. 1993, 58, (18), 4791-4793.