Thermo Scientific Chemicals

Benzyl acetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A14915.14
25 g, Each
Thermo Scientific Chemicals

Benzyl acetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A14915.14
25 g, Each
Quantity
Catalog number: A14915.14
also known as A14915-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5396-89-4
IUPAC Name
benzyl 3-oxobutanoate
Molecular Formula
C11H12O3
InChI Key
WOFAGNLBCJWEOE-UHFFFAOYSA-N
SMILES
CC(=O)CC(=O)OCC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5100-1.5150 @ 20?C

Description

Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

Solubility
It is soluble in alkali solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text