Thermo Scientific Chemicals

tert-Butyl bromoacetate, 98%, Thermo Scientific Chemicals

Catalog number: A14917.18
50 g, Each
Thermo Scientific Chemicals

tert-Butyl bromoacetate, 98%, Thermo Scientific Chemicals

Catalog number: A14917.18
50 g, Each
Quantity
Catalog number: A14917.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5292-43-3
IUPAC Name
tert-butyl 2-bromoacetate
Molecular Formula
C6H11BrO2
InChI Key
BNWCETAHAJSBFG-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)CBr
Identification (FTIR)
Conforms
Refractive Index
1.4430-1.4470 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥97.5%

Description

tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text