Thermo Scientific Chemicals

Tetra-n-octylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A14929.30
250 g, Each
Thermo Scientific Chemicals

Tetra-n-octylammonium bromide, 98+%, Thermo Scientific Chemicals

Catalog number: A14929.30
250 g, Each
Quantity
Catalog number: A14929.30
also known as A14929-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14866-33-2
IUPAC Name
tetraoctylazanium bromide
Molecular Formula
C32H68BrN
InChI Key
QBVXKDJEZKEASM-UHFFFAOYSA-M
SMILES
[Br-].CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC
Appearance (Color)
White to pale cream
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%
Melting Point (clear melt)
95-100?C
Form
Crystals or powder or crystalline powder or flakes
Identification (FTIR)
Conforms

Description

Tetra-n-octylammonium bromide is associated with benzethonium chloride and used as a disinfectant, in surface treatment and preservative in hospitals and industries. It is used as a surfactant involved in the synthesis of blue-luminescent silicon nanoparticles in order to get uniform particle size. It is utilized as a cationic additive in the preparation of membranes using cyclam derivative 1,4,8,11-tetra(n-octyl)-1,4,8,11-tetraazacyclotetradecane as an ionphore and dibutyl phthalate as plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-octylammonium bromide is associated with benzethonium chloride and used as a disinfectant, in surface treatment and preservative in hospitals and industries. It is used as a surfactant involved in the synthesis of blue-luminescent silicon nanoparticles in order to get uniform particle size. It is utilized as a cationic additive in the preparation of membranes using cyclam derivative 1,4,8,11-tetra(n-octyl)-1,4,8,11-tetraazacyclotetradecane as an ionphore and dibutyl phthalate as plasticizers.

Solubility
Soluble in water, dichloromethane, chloroform, acetone and methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text