Thermo Scientific Chemicals

Triphosgene, 98%, Thermo Scientific Chemicals

Catalog number: A14932.14
25 g, Each
Thermo Scientific Chemicals

Triphosgene, 98%, Thermo Scientific Chemicals

Catalog number: A14932.14
25 g, Each
Quantity
Catalog number: A14932.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32315-10-9
IUPAC Name
ditrichloromethyl carbonate
Molecular Formula
C3Cl6O3
InChI Key
UCPYLLCMEDAXFR-UHFFFAOYSA-N
SMILES
ClC(Cl)(Cl)OC(=O)OC(Cl)(Cl)Cl
Form
Crystals or powder or crystalline powder or lumps or fused solid
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to white to pale cream to pale yellow
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphosgene is used as a carbonylating agent for aza-peptide synthesis. It reacts with several alfa-amino acids to give the corresponding N-carboxyanhydrides. It is involved in the preparation of the esterification coupling reagent, di-2-thienyl carbonate from 2(5H)-thiophenone. Further, it is used as a reagent in organic synthesis and converts an amino group into isocyanate. In addition to this, it is employed in the preparation of 2-chloronicotinaldehydes through cyclization of the corresponding enamides. It is considered as a useful substitute for phosgene.

Solubility
Soluble in ether, ethanol, tetrahydrofuran, benzene, cyclohexane, chloroform, hexane, carbon tetrachloride, dichloromethane and chlorobenzene. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text