Thermo Scientific Chemicals

Potassium trihydrogen dioxalate dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A14933.30
250 g, Each
Thermo Scientific Chemicals

Potassium trihydrogen dioxalate dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A14933.30
250 g, Each
Quantity
Catalog number: A14933.30
also known as A14933-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6100-20-5
IUPAC Name
potassium oxalic acid hydrogen oxalate
Molecular Formula
C4H3KO8
InChI Key
GANDVAJEIJXBQJ-UHFFFAOYSA-M
SMILES
[K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
Form
Crystals or powder or crystalline powder
Assay (Redox Titration)
≥98.0 to ≤102.0%
Appearance (Color)
White
Identification (FTIR)
Conforms

Description

Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trihydrogen dioxalate dihydrat is used in metal polishing determination of pH. And it is also used in photography, marble grinding, and to remove ink stains.

Solubility
Soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text