2,6-Dimethoxy-p-benzoquinone, 98%, Thermo Scientific Chemicals
2,6-Dimethoxy-p-benzoquinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethoxy-p-benzoquinone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A14936.03
also known as A14936-03
Price (USD)/ Each
30.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
30.00
Add to cart
2,6-Dimethoxy-p-benzoquinone, 98%, Thermo Scientific Chemicals
Catalog numberA14936.03
Price (USD)/ Each
30.00
-
Add to cart
Chemical Identifiers
CAS530-55-2
IUPAC Name2,6-dimethoxycyclohexa-2,5-diene-1,4-dione
Molecular FormulaC8H8O4
InChI KeyOLBNOBQOQZRLMP-UHFFFAOYSA-N
SMILESCOC1=CC(=O)C=C(OC)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only