4-Chromanol, 97%, Thermo Scientific Chemicals
4-Chromanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chromanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A14940.06
also known as A14940-06
Price (USD)/ Each
97.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
97.90
Add to cart
4-Chromanol, 97%, Thermo Scientific Chemicals
Catalog numberA14940.06
Price (USD)/ Each
97.90
-
Add to cart
Chemical Identifiers
CAS1481-93-2
IUPAC Name3,4-dihydro-2H-1-benzopyran-4-ol
Molecular FormulaC9H10O2
InChI KeyMGSHXMOLUWTMGP-UHFFFAOYNA-N
SMILESOC1CCOC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)Pale cream or pale yellow
Melting Point (clear melt)37.0-43.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only