Thermo Scientific Chemicals

Methyl propionate, 99%, Thermo Scientific Chemicals

Catalog number: A14944.AE
100 mL, Each
Thermo Scientific Chemicals

Methyl propionate, 99%, Thermo Scientific Chemicals

Catalog number: A14944.AE
100 mL, Each
Quantity
Catalog number: A14944.AE
also known as A14944-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-12-1
IUPAC Name
methyl propanoate
Molecular Formula
C4H8O2
InChI Key
RJUFJBKOKNCXHH-UHFFFAOYSA-N
SMILES
CCC(=O)OC
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3750-1.3780 @ 20?C
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

Solubility
Soluble in water (5g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text